missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UNC5CL Monoclonal Antibody (3G7)

Product Code. 15523202
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15523202 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15523202 Supplier Invitrogen™ Supplier No. MA523197

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MCPQESSFQP SQFLLLVGVP VASVLLLAQC LRWHCPRRLL GACWTLNGQE EPVSQPTPQL ENEVSRQHLP ATLPEMVAFY QELHTPTQGQ TMVRQLMHKL LVFSAREVDH RGGCLMLQDT GISLLIPPGA VAVGRQERVS LILVWDLSDA PSLSQAQGLV SPVVACGPHG ASFLKPCTLT FKHCAEQPSH ARTYSSNTTL LDAKVWRPLG RPGAHASRDE CRIHLSHFSL YTCVLEAPVG REARKWLQLA VFCSPLVPGQ SHLQLRIYFL NNTPCALQWA LTNEQPHGGR LRGPCQLFDF NGARGDQCLK LTYISEGWEN VDDSSCQLVP HLHIWHGKCP FRSFCFRRKA ADENEDCSAL TNEIIVTMHT FQDGLETKYM EILRFQASEE ESWAAPPPVS QPPPCNRLPP ELFEQLRMLL EPNSITGNDW RGLASHLGLC GMKIRFLSCQ RSPAAAILEL FEEQNGSLQE LHYLMTVMER LDCASAIQNY LSGTHGGSPG PERGGARDNQ GLELDEKL.

UNC5CL is a membrane protein belonging to theUNC-5 family with a death domain and a ZU5 domain and is known to interact with p65/RELA and NFKB1. UNC5CL is known to interact with subunit p105 of NF-kappaB and transactivator p65 and is thus involved in inhibition of NF-kappaB activation and NF-kappaB-dependent transcription by inhibiting the binding of NF-kappaB to its target sequence (providing an alternative regulatory mechanism for NF-kappaB-mediated transcription). This regulatory protein sensitizes cells to apoptosis induced by TNF and TRAIL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5CL
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3G7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene UNC5CL
Gene Accession No. Q8IV45
Gene Alias 2510009H09Rik; BB155270; MGC34763; MUXA; OTTHUMP00000016362; protein unc-5 homolog C-like; unc-5 family C-terminal like; unc-5 homolog C (C. elegans)-like; UNC5CL; UNC5C-like protein; zu-5; ZU5 and death domain containing; ZU5 and death domain-containing inhibitor of NF-kB; ZU5 and death domain-containing protein; ZUD
Gene Symbols UNC5CL
Host Species Mouse
Immunogen UNC5CL (AAH35284, 1 a.a. ~ 518 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.