missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC45A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15479-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen.Especificaciones
| UNC45A | |
| Polyclonal | |
| Unconjugated | |
| UNC45A | |
| GC UNC45, GCUNC 45, GCUNC45, IRO039700, Protein unc 45 homolog A, SMAP 1, SMAP1, unc 45 homolog A (C. elegans), Unc 45A, UNC45A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101869, 308759, 55898 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q32PZ3, Q99KD5, Q9H3U1 | |
| UNC45A | |
| UNC45A Fusion Protein Ag7790 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido