missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC45A Antibody (AbS1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-89059
This item is not returnable.
View return policy
Description
UNC45A Monoclonal specifically detects UNC45A in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| UNC45A | |
| Monoclonal | |
| Unconjugated | |
| FLJ10043, GCUNC45, GC-UNC45, GCUNC-45, general cell UNC45, protein unc-45 homolog A, SMAP-1IRO039700, SMAP1UNC-45A, smooth muscle cell associated protein-1, Smooth muscle cell-associated protein 1, unc-45 homolog A (C. elegans), Unc-45A | |
| Recombinant human UNC45A. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| AbS1 | |
| PBS containing 50% glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 55898 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction