missing translation for 'onlineSavingsMsg'
Learn More

UNC2025, MedChemExpress

Product Code. 16417837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417837 5 mg 5mg
16485108 10 mg 10mg
16451347 50 mg 50mg
16435836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16417837 Supplier Medchem Express Supplier No. HY12344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC2025 is a potent, ATP-competitive and highly orally active Mer/Flt3 inhibitor with IC50 values of 0.74 nM and 0.8 nM, respectively. UNC2025 is >45-fold selectivity for MERTK relative to Axl (IC50= 122 nM; Ki = 13.3 nM). UNC2025 exhibits an excellent PK properties, and can be used for the investigation of acute leukemia.

UNC2025 is a potent, ATP-competitive and highly orally active Mer/Flt3 inhibitor with IC50 values of 0.74 nM and 0.8 nM, respectively. UNC2025 is >45-fold selectivity for MERTK relative to Axl (IC50= 122 nM

Specifications

Chemical Name or Material UNC2025
CAS 1429881-91-3
Color White
Purity Grade Notes Research
Molecular Formula C28H40N6O
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (69.92 mM; Need ultrasonic and warming)
SMILES CN1CCN(CC2=CC=C(C3=CN([C@@H]4CC[C@@H](O)CC4)C5=NC(NCCCC)=NC=C53)C=C2)CC1
Molecular Weight (g/mol) 476.66
Formula Weight 476.66
Percent Purity 96.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.