missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Unc18-2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16596791
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16596791 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16596791 Supplier Invitrogen™ Supplier No. PA588599

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAPSGLKAVV GEKILSGVIR SVKKDGEWKV LIMDHPSMRI LSSCCKMSDI LAEGITIVED INKRREPIPS LEAIYLLSPT EKALIKDFQG TPTFTYKAAH IFFTDTCPEP LFSELGRSRL AKVVKTLKEI HLAFLPYEAQ VFSLDAPHST YNLYCPFRAE ERTRQLEVLA QQIATLCATL QEYPAIRYRK GPEDTAQLAH AVLAKLNAFK ADTPSLGEGP EKTRSQLLIM DRAADPVSPL LHELTFQAMA YDLLDIEQDT YRYETTGLSE AREKAVLLDE; Positive Samples: THP-1, Jurkat, Mouse testis, Mouse thymus.

This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Unc18-2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STXBP2
Gene Accession No. Q15833, Q64324
Gene Alias C79054; FHL5; Hunc18b; Munc-18-2; Munc18-2; Munc18b; Munc-18b; muSec1; pp10122; Protein unc-18 homolog 2; protein unc-18 homolog B; Stxbp2; Sxtbp2; Sxtp2; syntaxin binding protein 2; syntaxin binding protein Munc18-2; syntaxin-binding protein 2; unc-18 homolog 2; unc-18 homolog B; Unc18-2; Unc18b; unc-18B
Gene Symbols STXBP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human STXBP2 (NP_0011208681).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20911, 6813
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.