missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Unc18-2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA576251

Product Code. 15962775

  • £472.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Unc18-2
Polyclonal
Unconjugated
STXBP2
C79054; FHL5; Hunc18b; Munc-18-2; Munc18-2; Munc18b; Munc-18b; muSec1; pp10122; Protein unc-18 homolog 2; protein unc-18 homolog B; Stxbp2; Sxtbp2; Sxtp2; syntaxin binding protein 2; syntaxin binding protein Munc18-2; syntaxin-binding protein 2; unc-18 homolog 2; unc-18 homolog B; Unc18-2; Unc18b; unc-18B
Rabbit
Antigen affinity chromatography
RUO
20911, 6813
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q15833, Q64324
STXBP2
Recombinant fusion protein of human Unc18-2.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis