missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Unc18-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550281
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total STXBP2 protein.
This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
| Unc18-2 | |
| Polyclonal | |
| Unconjugated | |
| STXBP2 | |
| C79054; FHL5; Hunc18b; Munc-18-2; Munc18-2; Munc18b; Munc-18b; muSec1; pp10122; Protein unc-18 homolog 2; protein unc-18 homolog B; Stxbp2; Sxtbp2; Sxtp2; syntaxin binding protein 2; syntaxin binding protein Munc18-2; syntaxin-binding protein 2; unc-18 homolog 2; unc-18 homolog B; Unc18-2; Unc18b; unc-18B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6813 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q15833 | |
| STXBP2 | |
| Fusion protein corresponding to residues near the C terminal of human Syntaxin binding protein 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction