missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC13B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49364-25ul
This item is not returnable.
View return policy
Description
UNC13B Polyclonal antibody specifically detects UNC13B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| UNC13B | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| hmunc13, homolog of rat Munc13 (diacylglycerol-binding), MGC133279, MGC133280, MUNC13, Munc13-2, protein unc-13 homolog B, unc-13 homolog B (C. elegans), unc-13-like (C. elegans), UNC13Unc13h2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PPDLVLQKDHFLGPQESFPEENASSPFTQARAHWIRAVTKVRLQLQEIPDD | |
| 25 μL | |
| Apoptosis | |
| 10497 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction