missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC13B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | UNC13B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676711
|
Novus Biologicals
NBP2-93337-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607051
|
Novus Biologicals
NBP2-93337-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UNC13B Polyclonal antibody specifically detects UNC13B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| UNC13B | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.3), 50% glycerol | |
| 10497 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| hmunc13, homolog of rat Munc13 (diacylglycerol-binding), MGC133279, MGC133280, MUNC13, Munc13-2, protein unc-13 homolog B, unc-13 homolog B (C. elegans), unc-13-like (C. elegans), UNC13Unc13h2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human UNC13B (NP_006368.3). MSLLCVRVKRAKFQGSPDKFNTYVTLKVQNVKSTTVAVRGDQPSWEQDFMFEISRLDLGLSVEVWNKGLIWDTMVGTVWIALKTIRQSDEEGPGEWSTLEAETLMKDDEICGTRNPTPHKILLDTRFELPFDIPEEEARYWTYKWEQINALGADNEYSSQEESQRKPLPTAAAQCSFEDPDSAVDDRDSDYRSETSNSFPPPYHTASQPNASVHQFPVPVRSPQQLLLQGSSRDSCNDSMQSYDLDYPERRAISPTSSSRYGSSCNVSQGSSQLSELDQYHEQDDDHRETDSIHSCHSSHSLSRDGQAGFGEQEKPLEVTGQAEKEAACEPKEMKEDATTHPPPDLVLQK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title