missing translation for 'onlineSavingsMsg'
Learn More

UNC1215, MedChemExpress

Product Code. 16492797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492797 50 mg 50mg
16489267 5 mg 5mg
16476718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492797 Supplier Medchem Express Supplier No. HY15649
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress UNC1215 is a potent and selective inhibitor for the methyllysine (Kme) reading domain function of L3MBTL3 with a Kd value of 120 nM and an IC50 of 40 nM. UNC1215 has the potential to treat malignant brain tumor.

UNC1215 is a potent and selective inhibitor for the methyllysine (Kme) reading domain function of L3MBTL3 with a Kd value of 120 nM and an IC50 of 40 nM. UNC1215 has the potential to treat malignant brain tumor.

Specifications

Chemical Name or Material UNC1215
CAS 1415800-43-9
Color White
Purity Grade Notes Research
Molecular Formula C32H43N5O2
Quantity 50 mg
Solubility Information DMSO : ≥ 270 mg/mL (509.70 mM)
SMILES O=C(C1=CC=C(C(N2CCC(N3CCCC3)CC2)=O)C=C1NC4=CC=CC=C4)N5CCC(N6CCCC6)CC5
Molecular Weight (g/mol) 529.72
Formula Weight 529.72
Percent Purity 98.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.