missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC119 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20811
This item is not returnable.
View return policy
Description
UNC119 Polyclonal specifically detects UNC119 in Human samples. It is validated for Western Blot.
Specifications
| UNC119 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| hRG4, HRG4POC7 centriolar protein homolog A, POC7, POC7A, protein unc-119 homolog A, Retinal protein 4, RG4, unc119 (C.elegans) homolog, unc-119 homolog (C. elegans) | |
| Rabbit | |
| 27 kDa | |
| 0.1 mL | |
| Vision | |
| 9094 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q13432 | |
| UNC119 | |
| Recombinant protein encompassing a sequence within the center region of human UNC119. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction