missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UNC 6852
GREENER_CHOICE

Product Code. 18795805
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18795805

Brand: Tocris Bioscience™ 7816/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polycomb repressive complex 2 (PRC2) Degrader PROTAC™

UNC 6852 is a polycomb repressive complex 2 (PRC2) Degrader (PROTAC™; DC50 = 0.79 μM). It comprises an EED (embryonic ectoderm development) based ligand, joined by a linker to a von Hippel-Lindau ligand. In B lymphoblast cell lines, UNC 6852 selectively degrades PRC2 components EED (DC50 = 0.31 μM, Dmax = 94%), enhancer of zeste homolog 2 (EZH2, DC50 = 0.67 μM, Dmax = 96%), and suppressor of zeste homolog 12 (SUZ12, DC50 = 0.59 μM, Dmax = 82%). In zebrafish, UNC 6852 treatment increases transcription of tfpia and prolongs time to occlusion (TTO) in the caudal vein following laser injury.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-(4-(4-(5-((Furan-2-ylmethyl)amino)-[1,2,4 ]triazolo[4,3-c]pyrimidin-8-yl)benzamido)butanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2688842-08-0
Quantity 5 mg
Target Active Degraders
Molecular Formula C43H48N10O6S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.