missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UNC 3230
GREENER_CHOICE

Product Code. 18786119
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18786119

Brand: Tocris Bioscience™ 5271/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PIP5K1C inhibitor; antinociceptive

UNC 3230 is a potent and selective PIP5K1C inhibitor (IC50 = 41 nM). Exhibits selectivity for PIP5K1C over PIP5K1A, the PI 3-kinase family and a panel of other kinases. Reduces PIP2 levels and LPA-induced calcium signaling in dorsal root ganglia (DRG) neurons in vitro. Reduces nociception in mouse models of chronic pain.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type UNC 3230
Purity >99%
Molecular Weight (g/mol) 344.43
Inhibitors UNC 3230

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.