missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UNC 1215, acid
GREENER_CHOICE

Product Code. 18768986
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18768986

Brand: Tocris Bioscience™ 7759/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Functionalized L3MBTL3 ligand for PROTACs

UNC 1215, acid is a functionalized L3MBTL3 ligand for PROTAC® research and development. L3MBTL3-recruiting PROTACs induce nuclear-specific protein degradation of target proteins, such as FKBP12(F36V) and BRD2. UNC 1215, acid contains a carboxylic acid handle ready for conjugation to a target protein ligand.

Part of a range of functionalized tool molecules for PROTAC® Degrader R&D.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[[2,5-Bis[[4-(1-pyrrolidinyl)-1-piperidinyl]carbonyl]phenyl]amino]benzoic acid ditrifluoroacetate
Quantity 50 mg
Target Degrader Building Blocks
Molecular Formula C33H43N5O4.2CF3CO2H
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.