missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 0638

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective inhibitor of G9a and GLP histone lysine methyltransferases (IC50 values are < 15 nM and 19 nM for G9a and GLP respectively, and > 10000 nM for a range of other histone methyltransferases). Potently inhibits dimethylation of H3K9 in MCF-7 cells (IC50 = 70 nM). Cell permeable.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 509.73 |
| Purity | >99% |
| Molecular Weight (g/mol) | 527.754 |
| Chemical Name or Material | UNC 0638 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction