missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UMPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557560
This item is not returnable.
View return policy
Description
Immunogen sequence: RLLSQVADIL FQTAQNAGIS FDTVCGVPYT ALPLATVICS TNQIPMLIRR KETKDYGTKR LVEGTINPGE TCLIIEDVVT SGSSVL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a uridine 5'-monophosphate synthase. The encoded protein is a bifunctional enzyme that catalyzes the final two steps of the de novo pyrimidine biosynthetic pathway. The first reaction is carried out by the N-terminal enzyme orotate phosphoribosyltransferase which converts orotic acid to orotidine-5'-monophosphate. The terminal reaction is carried out by the C-terminal enzyme OMP decarboxylase which converts orotidine-5'-monophosphate to uridine monophosphate. Defects in this gene are the cause of hereditary orotic aciduria. Alternate splicing results in multiple transcript variants.
Specifications
| UMPS | |
| Polyclonal | |
| Unconjugated | |
| UMPS | |
| 1700095D23Rik; AA408257; AL033308; BB164745; mutant uridine monophosphate synthase; ODC; OK/SW-cl.21; OMPdecase; OPRT; OPRTase; orotate phosphoribosyl transferase and orotidine-5'-decarboxylase; orotate phosphoribosyltransferase; Orotidine 5'-phosphate decarboxylase; UMP synthase; UMPS; Uridine 5'-monophosphate synthase; uridine 5-monophosphate synthase; uridine 5'-monophosphate synthase-like protein; uridine monophophate synthase; uridine monophosphate synthase; uridine monophosphate synthetase; uridine monophosphate synthetase (orotate phosphoribosyl transferase and orotidine-5'-decarboxylase); uridine monophosphate synthetase [orotate phosphoribosyl transferase and orotidine-5'-decarboxylase] | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22247, 288051, 7372 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11172, P13439 | |
| UMPS | |
| Recombinant protein corresponding to Human UMPS. Recombinant protein control fragment (Product #RP-96306). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction