missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UMPS Polyclonal antibody specifically detects UMPS in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 430 to 480 of human Uridine Monophosphate Synthetase using the numbering given in entry NP_000364.1 (GeneID 7372). |
| Purification Method | Immunogen affinity purified |
| Regulatory Status | RUO |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?