missing translation for 'onlineSavingsMsg'
Learn More

UMI-77, MedChemExpress

Product Code. 16410028
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16410028

Brand: Medchem Express HY18628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UMI-77 is a selective Mcl-1 inhibitor, which shows high binding affinity to Mcl-1 (IC50=0.31 μM). UMI-77 binds to the BH3 binding groove of Mcl-1 with Ki of 490 nM, showing selectivity over other members of anti-apoptotic Bcl-2 members.

UMI-77 is a selective Mcl-1 inhibitor, which shows high binding affinity to Mcl-1 (IC50=0.31 μM). UMI-77 binds to the BH3 binding groove of Mcl-1 with Ki of 490 nM, showing selectivity over other members of anti-apoptotic Bcl-2 members.

Specifications

Chemical Name or Material UMI-77
CAS 518303-20-3
Color White
Purity Grade Notes Research
Molecular Formula C18H14BrNO5S2
Quantity 5 mg
Solubility Information DMSO : ≥ 28 mg/mL (59.79 mM)
SMILES O=C(O)CSC1=CC(NS(=O)(C2=CC=C(Br)C=C2)=O)=C3C=CC=CC3=C1O
Molecular Weight (g/mol) 468.34
Formula Weight 468.34
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.