Learn More
Description
- Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
- Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
- Reduced cost per test — Quanto remains one of the most affordable systems available
- Shorter incubation times — Faster turn around times
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: None provided
Includes: TL-060-HL: 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB)
TL-125-HL: 125mL Primary Antibody Amplifier Quanto (TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB),
Specifications
Specifications
| Quantity | 60 mL |
| Includes | 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB) |
| Chromagen | None provided |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Storage Requirements | Store at 2° to 8°C. Each component is stable for up to 18 months. |
| Enzyme | Peroxidase |
| Product Line | UltraVision |
| Substrate | None provided |
Safety and Handling
- UltraVision Quanto Detection
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: biphenyl-3,3',4,4'-tetrayltetraamine, diaminobenzidine, 2-methyl-3(2H)-isothiazolone, Sodium azide
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.