missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraVision™ Quanto Detection System HRP

Product Code. 12694007 Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
Unit Size:
125mL
60mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12694007 60 mL 60mL
12684017 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12694007 Supplier Epredia™ Supplier No. TL060QHL

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Epredia™ UltraVision Quanto HRP DAB delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.

UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.
  • Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
  • Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
  • Reduced cost per test — Quanto remains one of the most affordable systems available
  • Shorter incubation times — Faster turn around times
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes: TL-060-HL: 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB)

TL-125-HL: 125mL Primary Antibody Amplifier Quanto (TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB),

TRUSTED_SUSTAINABILITY

Specifications

Quantity 60 mL
Includes 60mL Primary Antibody Amplifier Quanto (TL-060-QPB), 60mL HRP Polymer Quanto (TL-060-QPH), 60mL Ultra V Block (TA-060-UB)
Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Each component is stable for up to 18 months.
Enzyme Peroxidase
Product Line UltraVision
Substrate None provided
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • UltraVision Quanto Detection
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: biphenyl-3,3',4,4'-tetrayltetraamine, diaminobenzidine, 2-methyl-3(2H)-isothiazolone, Sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.