missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraVision™ Quanto Detection System HRP DAB

Product Code. 12673997 Shop All Epredia Products
Change view
Click to view available options
Quantity:
15 mL
60 mL
125 mL
Unit Size:
125mL
15mL
60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12673997 15 mL 15mL
12664007 60 mL 60mL
12674017 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12673997 Supplier Epredia™ Supplier No. TL015QHD
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Epredia™ UltraVision Quanto HRP DAB delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.

UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.
  • Enhanced sensitivity while minimizing background — Can be used with low expression and routine markers alike
  • Increased sensitivity for rabbit antibodies — more economical use of expensive rabbit monoclonal antibodies
  • Reduced cost per test — Quanto remains one of the most affordable systems available
  • Shorter incubation times — Faster run times

Includes: TL-060-QHD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Primary Antibody Amplifier Quanto(TL-060-QPB), 60mL HRP Polymer Quanto(TL-060-QPH), 60mL Ultra V Block (TA-060-UB), 60mL DAB Quanto Substrate (TA-060-QHSX), 2mL DAB Quanto Chromogen (TA-002-QHCX)

TL-125-QHD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Primary Antibody Amplifier Quanto(TL-125-QPB), 125mL HRP Polymer Quanto (TL-125-QPH), 125mL Ultra V Block (TA-125-UB), 125mL DAB Quanto Substrate (TA-125-QHSX), 4mL DAB Quanto Chromogen (TA-004-QHCX)

TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 mL
Includes 15mL Hydrogen Peroxide,15mL Antibody Amplifier Quanto,15mL HRP Polymer, 15mL UltraV Block, 15mL DAB Quanto Substrate, DAB Quanto Chromogen
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 15 ML ULTRAVISION QUANTO DETECTION SYSTEM HRP DAB, contains 15mL Hydrogen Peroxide,15mL Antibody Amplifier Quanto,15mLHRP Polymer, 15mL UltraV Block, 15mL DAB Quanto Substrate, DAB Quanto Chromogen 15mL
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P321-Specific treatment (see on this label).
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3,3'-Diaminobenzidine tetrahydrochloride, 2-methyl-3(2H)-isothiazolone, triton-X100, sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.