missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraVision Quanto Detection System AP

Product Code. 12664017 Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
Substrate:
Diaminobenzidine (DAB)
None provided
Enzyme:
Alkaline Phosphatase
Peroxidase
Unit Size:
125mL
60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Substrate Enzyme Unit Size
12664017 125 mL None provided Alkaline Phosphatase 125mL
12654007 60 mL None provided Alkaline Phosphatase 60mL
15290715 125 mL Diaminobenzidine (DAB) Peroxidase 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12664017 Supplier Epredia™ Supplier No. TL125QAL
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.

UltraVision Quanto AP has been validated in combination with Liquid Fast-Red Substrate System and provides four major benefits:

High sensitivity—can both be used for tricky and challenging markers and/or allows a more gentle HIER

Increased sensitivity for rabbit antibodies—more economical use of expensive rabbit monoclonal antibodies Crisp, clear and strong staining for high contrast

Reduced incubation times for shorter run times&bulletc1_end; UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Alkaline Phosphatase

  • Chromogen/Substrate: None provided

Includes: TL-060-AL: Primary Antibody Amplifier Quanto (TL-060-QPB), AP Polymer Quanto (TL-060-QAP), Ultra V Block (TA-060-UB)

TL-125-AL: Primary Antibody Amplifier Quanto (TL-125-QPB), AP Polymer Quanto (TL-125-QAP), Ultra V Block (TA-125-UB)

TRUSTED_SUSTAINABILITY

Specifications

Chromagen None provided
Enzyme Alkaline Phosphatase
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Substrate None provided
Includes Primary Antibody Amplifier Quanto (TL-125-QPB), AP Polymer Quanto (TL-125-QAP), Ultra V Block (TA-125-UB)
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.