missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraVision LP HRP Polymer & DAB Detection System

Product Code. 12624017 Shop All Epredia Products
Click to view available options
Quantity:
15 mL
60 mL
125 mL
Unit Size:
125mL
15mL
60mL
This item is not returnable. View return policy

Product Code. 12624017

Brand: Epredia™ TL125HD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Epredia™ Lab Vision UltraVision LP Detection System: HRP Polymer/DAB Plus Chromogen is a sensitive detection system based on biotin-free, polymer technology.

UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification.1 Ultimately, this gives the user higher sensitivity and antibody efficiency.2 With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: Diaminobenzidine (DAB)

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: Diaminobenzidine (DAB)

Includes:

TL-015-HD: 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)

TL-060-HD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH), 60mL DAB Plus Substrate (TA-060-HSX), 2mL DAB Plus Chromogen (TA-002-HCX)

TL-125-HD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)

Note:

UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.

TRUSTED_SUSTAINABILITY

Specifications

Count 625 to 1250 slides
Quantity 125 mL
Includes 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)
Chromagen Diaminobenzidine (DAB)
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Each component is stable for 18 months.
Enzyme Peroxidase
Product Line Lab Vision, UltraVision
Substrate Diaminobenzidine (DAB)
compliance-icons compliance-icons
Product Identifier
  • 125 ML ULTRAVISION LP DETECTION SYSTEM: HRP POLYME r/DAB Plus chromogen, specificity anti-Mouse IgG
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : biphenyl-3,3',4,4'-tetrayltetraamine, diaminobenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.