Learn More
Excel Healthcare Ltd™ Ultrasonic Cleaner
ultrasonic cleaning enhances the removal of debris prior to sterilisation
Brand:  Excel Healthcare Ltd™ 285
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 16300132
  • POA / Each
Estimated Shipment: 02-07-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Quite Ultrasonic Noise Frequency
  • Lcd Display
  • Heated Element
  • Time/Heating Adjustment
  • Light Weight And Compact Unit
  • Ultrasonic Cleaning Enhances The Removal Of Debris Prior To Sterilisation
  • It Is Vital That The Water And Detergent Are Well Maintained, Cleaned And Changed At Regular Intervals To Ensure Maximum Cleaning Effectiveness
  • Htm 01-05 Recommends That Intruments Placed In Ultrasonic Baskets Must Be Fully Immersed In The Cleaning Solution
  • After The Process Has Taken Place, Instruments Must Be Thoroughly Cleaned And Rinsed Using Distilled Water In Order To Remove Residual Soil And Detergents With Minimum Risk Of Salt Deposition
Specifications
Specifications
Digital Timer
31 x 37 x 30 cm
285
10 L
Plastic Basket, Stainless Steel Basket, Drain Hose, Power Lead