missing translation for 'onlineSavingsMsg'
Learn More

MICROFLEX™ UltraSense™ Nitrile Exam Gloves

Product Code. 12970745
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
12970745 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12970745 Supplier MICROFLEX™ Supplier No. 220S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance ASTM D 6319
Color Blue
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Sterility Non-sterile
For Use With (Application) Healthcare, dental, industrial safety, laboratory, EMS
Glove Cuff Style Beaded/Rolled
Quantity 100/Pk.
Tensile Strength 30 to 25 MPa
Length (Metric) 24.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.