missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UltraScence Pico Ultra Western Substrate
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
The UltraScence Pico Ultra Western Substrate, as a luminol-based enhanced chemiluminescent substrate, is sensitive and compatible with conducting immunoblots with horseradish peroxidase (HRP) - conjugated secondary antibodies. The low picogram to mid femtogram detection of antigen is enabled by UltraScence Pico Ultra Western Substrate's excellent sensitivity and long signal duration. Further, its long chemiluminescent signal duration makes both digital and film-based imaging possible without any loss of the signal. Appropriate primary and secondary antibody dilutions are suggested for attaining optimal signal intensity and duration.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Quantity | 100 mL |
| Content And Storage | Store at 4°C. Do not freeze. |
| Target | UltraScence Pico Ultra Western Substrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction