missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ UltraPure™ SSPE, 20X

Product Code. 17142949
Click to view available options
Quantity:
10 x 1 L
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17142949

Brand: Thermo Scientific™ 15591050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

UltraPure 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications.

  • Supplied in 1 L or in 10 x 1 L plastic bottles, sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2 M NaH2PO4, and 0.02 M EDTA at pH 7.4.
  • Performance and quality testing: no DNase or RNase activity detected
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Physical Form Liquid
Product Line UltraPure™
Quantity 10 x 1 L

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.