missing translation for 'onlineSavingsMsg'
Learn More

Ultramark 1621, TRC

Product Code. 30449659
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30449659 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30449659

Brand: Toronto Research Chemicals TRCU7009005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ultramark 1621
CAS 105809-15-2
Quantity 5 g
InChI Formula InChI=1S/C6H18N3P3/c1-10(2)7-11(3,4)9-12(5,6)8-10/h1-6H3/i1D,2D,3D,4D,5D,6D
IUPAC Name 2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2λ^{5},4λ^{5},6λ^{5}-triphosphacyclohexa-1,3,5-triene
Formula Weight 231.109
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.