missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ultra Streptavidin-HRP

Product Code. 10721565 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10721565 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10721565 Supplier Thermo Scientific™ Supplier No. N504
  • ONLINE EXCLUSIVE PRICE
    £161.00
    £149.65 / 1.5mL
    Save £11.35
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra Streptavidin-HRP

Streptavidin-HRP consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme. Streptavidin binds to biotin and the conjugated HRP provides a high level of activity for detection using an appropriate substrate system.

Features of Streptavidin-HRP:

  • Product Number: N504
  • Size: 1.5 mL
  • Formulation: Conjugate in stabilizer solution containing protein and preserved with 0.02% methyliso-thiazolone, 0.02% bromonitrodioxane, and 0.002% other active isothiazolones.
  • Storage: Shipped on cold packs. Store at 2-8°C. Stable until expiration date if stored under these conditions.
  • Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:400 - 1:2,000 in ELISA diluent (e.g., PBS containing 4% protease-free BSA) . The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Conjugate HRP
    Quantity 1.5 mL
    Form Liquid
    Product Type Streptavidin Conjugate
    Target Molecule Biotin
    Label or Dye HRP (Horseradish Peroxidase)
    compliance-icons
    Product Identifier
    • Ultra Streptavidin-HRP
    Signal Word
    • Warning
    Hazard Category
    • Skin sensitiser Category 1
    Hazard Statement
    • H317-May cause an allergic skin reaction.
    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contain: 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.