missing translation for 'onlineSavingsMsg'
Learn More

Ultra Low Particle V-Solv+Co Solvent

Product Code. 30312998
Change view
Click to view available options
Quantity:
5 gal.
Unit Size:
5 GAL
Product Code. Quantity unitSize
30312998 5 gal. 5 GAL
1 options
This item is not returnable. View return policy

Product Code. 30312998

Brand: VHG Labs VHGVSOLV+COULP5GAL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cobalt
CAS 7440-48-4
Analyte or Component Names Cobalt
Shipping Condition Room Temperature
Molecular Formula Co
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Co
Solution Type Single Solution
SMILES [Co]
Element Co
IUPAC Name Cobalt
Solvent or Matrix V-SOLV™ ICP Solvent
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 58
Quantity 5 gal.
Formula Weight 58.9332
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.