missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Ultra High Sensitivity ECL Kit

Product Code. 16468946
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468946 100 mL 100mL
16442336 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468946 Supplier MedChemExpress Supplier No. HYK1005
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MCE Ultra High Sensitivity ECL Kit is an ultra-sensitive, luminol-based enhanced chemiluminescent substrate for detecting horseradish peroxidase (HRP) labeled antibodies on western blots.

Specifications

Product Type Kits
Quantity 100 mL
Content And Storage Stored at 4°C, and is stable for 1 year. Protect from light.
Shipping Condition Room temperature
Target VD/VDR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.