missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Ultra Durable Floor Marking Tape 971
Description
- Excellent abrasion resistance
- Withstands lifting from forklift traffic
- Bright colors for high visibility
- Resists moisture and many chemicals
- Sticks on contact and stays where you put it
- Promotes clean removal without solvents and scraping
- Available in a variety of colours
Specifications
Specifications
| Dimensions (L x W) | 51 mm x 33 m |
| Length (Metric) | 32.9 m |
| Width (Metric) | 50.8 mm |
| Color | White |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?