missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Ultra Adhesive (UA) Quality Control Labels 95 x 95mm

Product Code. 15849481 Shop All Sampling Systems Products
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
Each
Pack of 500
This item is not returnable. View return policy

Product Code. 15849481

Brand: Sampling Systems™ 8220A03

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specially constructed high quality labels with an ultra adhesive foil backing. Sampling Systems™ Ultra Adhesive (UA) Quality Control Labels 95 x 95mm are ideal for use on sacks, drums, kegs, boxes, bags, and more.

  • Very high strength adhesive - will even stick on very dusty surfaces
  • Large writing area
  • Each roll individually packed
  • Ideal for sealing dusty sacks
  • The label will not only record the fact that the sack has been sampled but it will prevent the sack from leaking powder as it is handled
  • Even after being submerged in water for 48 hours the UA label was intact, it was securely attached to the bag and could be written on using a ball point pen
  • Suitable for sticking on almost any surface

Specifications

Description Ultra Adhesive (UA) - High strength foil backed paper labels
Color Yellow
Material Paper
Product Type Ultra Adhesive Label
Display Text Quality Control
Labels per Sheet 500/Roll
No. per Pack 500/Pk.
Temperature Range (Metric) -20°C to +70°C
Dimensions (LxW) 95 x 95 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.