missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK4 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41244AF594
This item is not returnable.
View return policy
Description
ULK4 Polyclonal antibody specifically detects ULK4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ULK4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| DKFZp434E1822, EC 2.7.11.1, FAM7C1, FLJ20574, REC01035, serine/threonine-protein kinase ULK4, Unc-51-like kinase 4, unc-51-like kinase 4 (C. elegans) | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human ULK4. The immunogen is located within amino acids 1120 - 1170 of ULK4. Amino Acid Squence: AQKSGSGEDPQAAEDL | |
| 0.1 mL | |
| Protein Kinase | |
| 54986 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction