missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULK3 Recombinant Rabbit Monoclonal Antibody (K01_2J84)
GREENER_CHOICE

Product Code. 30827888
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827888 100 μL
30827331 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827888 Supplier Invitrogen™ Supplier No. MA563916
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ULK3 Recombinant Monoclonal Antibody for Western Blot

ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2J84
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ULK3
Gene Accession No. Q6PHR2
Gene Alias 1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3
Gene Symbols ULK3
Host Species Rabbit
Immunogen A synthetic peptide of human ULK3.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25989
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.