missing translation for 'onlineSavingsMsg'
Learn More

ULK3 Antibody, Novus Biologicals™

Product Code. 18373589 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373589 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373589 Supplier Novus Biologicals Supplier No. H00025989B03P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ULK3 Polyclonal antibody specifically detects ULK3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Alias DKFZP434C131, EC 2.7.11.1, FLJ90566, serine/threonine-protein kinase ULK3, Unc-51-like kinase 3, unc-51-like kinase 3 (C. elegans)
Host Species Mouse
Immunogen ULK3 (AAH36117.1, 1 a.a. - 214 a.a.) full-length human protein. MAGPGWGPPRLDGFILTERLGSGTYATVYKAYAKKDTREVVAIKCVAKKSLNKASVENLLTEIEILKGIRHPHIVQLKDFQWDSDNIYLIMEFCAGGDLSRFIHTRRILPEKVARVFMQQLASALQFLHERNISHLDLKPQNILLSSLEKPHLKLADFGFAQHMSPWDEKHVLRGSPLYMAPEMVCQRQYDARVDLWSMGVILYGETSFPCFSP
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 25989
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.