missing translation for 'onlineSavingsMsg'
Learn More

ULK3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£134.00 - £398.00

Specifications

Antigen ULK3
Concentration 1 mg/mL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18676807
View Documents
Novus Biologicals
NBP2-41243-0.025mg
0.025 mg
£134.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18176359
View Documents
Novus Biologicals
NBP2-41243
0.1 mg
£398.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ULK3 Polyclonal specifically detects ULK3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

ULK3
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
25989
Antibody was raised against a 19 amino acid peptide near the center of human ULK3. The immunogen is located within amino acids 230 - 280 of ULK3. Amino Acid Squence: DLLQRLLERDPSRRISFQD
Primary
ULK3 antibody is human specific. Multiple isoforms of ULK3 are known to exist.
1 mg/mL
Polyclonal
Rabbit
Protein Kinase
PBS with 0.02% Sodium Azide
ULK3
IgG
Affinity Purified
52 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.