missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ULK2 Recombinant Rabbit Monoclonal Antibody (HL2206)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550432
This item is not returnable.
View return policy
Description
ULK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
ULK2, also known as ATG1B, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK2 results in a severe defect in the autophagy pathway. ULK2 is highly conserved among eukaryotes and shows high homology with its related protein ULK1. Both ULK1 and ULK2 form a complex with ATG13 and FIP200 that mediates TOR signaling and is essential for autophagy. Like ULK1, ULK2 is also thought to be involved in early neuronal growth and differentiation.
Specifications
| ULK2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8IYT8 | |
| ULK2 | |
| Recombinant fragmemt of human ULK2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2206 | |
| Unconjugated | |
| ULK2 | |
| A830085I22Rik; ATG1B; AU015340; EC:2.7.11.1; KIAA0623; mKIAA0623; serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; ULK2; unc-51 like autophagy activating kinase 2; unc-51 like kinase 2; Unc51.2; unc-51-like kinase 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 303206, 9706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction