missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-33136
This item is not returnable.
View return policy
Description
ULK2 Polyclonal specifically detects ULK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ULK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| ULK2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human ULK2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| ATG1B, EC 2.7.11, EC 2.7.11.1, KIAA0623serine/threonine-protein kinase ULK2, unc-51 (C. elegans)-like kinase 2, Unc51.2, Unc-51-like kinase 2, unc-51-like kinase 2 (C. elegans) | |
| Rabbit | |
| 113 kDa | |
| 100 μL | |
| Protein Kinase | |
| 9706 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction