missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ULK1, GST Tag Recombinant Protein

Product Code. p-7121941
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 15431234

Brand: Invitrogen™ PV6430

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human ULK1 (1-649) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Fragment Assay Entry: Biochemical binding/kinetic The gene accession number for ULK1 is NP_003556.1.

ULK1 is a serine/threonine protein kinase that plays critical role during initial stages of autophagy which is a vital response to nutrient starvation. The conserved C-terminal domain (CTD) of ULK1 controls the regulatory function and localization of the protein. Knockdown of ULK1 inhibits the autophagic response as well as inhibiting rapamycin-induced autophagy consistent with a role downstream of mTOR. ULK1 forms a complex with FIP200 and ATG13 and this complex is essential for starvation-induced autophagy. Both FIP200 and ATG13 are critical for correct localization of ULK1 to the pre-autophagosome and stability of ULK1 protein. ULK1 is phosphorylated by the mTOR pathway in a nutrient starvation-regulated manner.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75385
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.25mM DTT, 0.1mM EDTA, 0.1mM PMSF, 10mM glutathione, 25% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 8408
Molecular Weight (g/mol) 96.1 kDa
Name Human ULK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase Unc51.1; ULK 1; Ulk1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; unc-51-like kinase 1
Common Name ULK1
Gene Symbol ULK1
Protein Length 1-649
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-649
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic
Kinase Group Serine/Threonine Kinase
Target Entry ULK1
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Fragment
Form Liquid
Protein Subtype Serine/Threonine Kinases
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.