missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20986-1-AP-150UL
This item is not returnable.
View return policy
Description
ULK1, also known as ATG1, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK1 results in a severe defect in the autophagy pathway. ULK1 is highly conserved among eukaryotes, and are the Unc-51-like kinases, ULK1 and ULK2 in mammals. ULK1 is ubiquitously expressed and involved in autophagy in response to starvation. It is the target of the TOR kinase signaling pathway that regulates autophagy through the control of phosphorylation status of ATG13. ULK1 also plays a role early in neuronal differentiation.Specifications
| ULK1 | |
| Polyclonal | |
| Unconjugated | |
| ULK1 | |
| ATG1, KIAA0722, ULK1, Unc 51 like kinase 1, UNC51, Unc51.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360827, 8408 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75385 | |
| Ulk1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction