missing translation for 'onlineSavingsMsg'
Learn More

Ulipristal-d3, TRC

Product Code. 30455444
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30455444

Brand: Toronto Research Chemicals TRCU70080210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ulipristal-D3 (N-methyl-D3)
Molecular Formula C28 2H3 H32 N O3
Quantity 10 mg
Synonym (11β)-11-[4-[(Methyl)(methyl-d3)amino]phenyl]-17-hydroxy-19-norpregna-4,9-diene-3,20-dione, 11β-[4-(N-Methyl-N-(methyl-d3)amino)phenyl]-17-hydroxy-19-norpregna-4,9-diene-3,20-dione, Ulipristal-d3, Ulipristal-D3 (N-methyl-D3)
InChI Formula InChI=1S/C28H35NO3/c1-17(30)28(32)14-13-25-23-11-7-19-15-21(31)10-12-22(19)26(23)24(16-27(25,28)2)18-5-8-20(9-6-18)29(3)4/h5-6,8-9,15,23-25,32H,7,10-14,16H2,1-4H3/t23-,24+,25-,27-,28-/m0/s1/i3D3
SMILES [2H]C([2H])([2H])N(C)c1ccc(cc1)[C@H]2C[C@@]3(C)[C@@H](CC[C@]3(O)C(=O)C)[C@@H]4CCC5=CC(=O)CCC5=C24
IUPAC Name (8S,11R,13S,14S,17R)-17-acetyl-17-hydroxy-13-methyl-11-[4-[methyl(trideuteriomethyl)amino]phenyl]-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 436.601
Formula Weight 436.281
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.