missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ulipristal acetate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Ulipristal acetate (CDB-2914) is an orally active, selective progesterone receptor modulator (SPRM). Ulipristal acetate stimulates the autophagic response selectively in leiomyoma cells. Ulipristal acetate has the potential for benign gynecological conditions treatment, such as uterine myoma.
Specifications
Specifications
| Chemical Name or Material | Ulipristal acetate |
| Physical Form | Solid |
| CAS | 126784-99-4 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C30H37NO4 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | CDB-2914 |
| Solubility Information | DMSO : 33.33 mg/mL (70.08 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction