missing translation for 'onlineSavingsMsg'
Learn More

ULBP-6/RAET1L Antibody, DyLight 594, Novus Biologicals™

Product Code. 18377512 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377512 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377512 Supplier Novus Biologicals Supplier No. NBP306628DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ULBP-6/RAET1L Polyclonal antibody specifically detects ULBP-6/RAET1L in Human samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP-6/RAET1L
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias retinoic acid early transcript 1L protein, retinoic acid early transcript 1L, ULBP6
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ULBP-6/RAET1L (AAK91503.1; Met1-Ser217).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 154064
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.