missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-4/RAET1E Antibody (RAET1E 119/6), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50266PECY7
This item is not returnable.
View return policy
Description
ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Flow Cytometry.
Specifications
| ULBP-4/RAET1E | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| RAET1E | |
| Recombinant His-tagged protein to human extra-cellular domain of ULBP-4/RAET1E produced in E.coli (33-227) with a N terminal 6x His tag. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| RAET1E 119/6 | |
| Flow Cytometry | |
| bA350J20.7, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E2, retinoic acid early transcript 1ERL-4, ULBP4MGC125308 | |
| Mouse | |
| Protein A purified | |
| Immunology | |
| 135250 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction