missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ULBP-2 Polyclonal antibody specifically detects ULBP-2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | ULBP-2 |
| Classification | Polyclonal |
| Dilution | ELISA |
| Gene Alias | ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human ULBP-2 (Uniprot#: Q9BZM5; Met1-Ser217) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?