missing translation for 'onlineSavingsMsg'
Learn More

ULBP-2 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18351002 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351002 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351002 Supplier Novus Biologicals Supplier No. NBP299939G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ULBP-2 Polyclonal specifically detects ULBP-2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP-2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ULBP-2 (Q9BZM5; Met 1-Ser 217).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.