missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-2 Antibody (003), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90059C
This item is not returnable.
View return policy
Description
ULBP-2 Monoclonal specifically detects ULBP-2 in Human samples. It is validated for ELISA.
Specifications
| ULBP-2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 80328 | |
| Human | |
| Purified |
| ELISA | |
| 3 | |
| ELISA | |
| ALCAN-alpha, N2DL2, N2DL-2, NKG2D ligand 2, RAET1HNKG2DL2, retinoic acid early transcript 1 H, Retinoic acid early transcript 1H, UL16 binding protein 2, UL16-binding protein 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ULBP-2 (Q9BZM5; Met1-Ser217). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human ULBP1 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction