missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-1 Antibody (MM0592-10K10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £423.00
Specifications
| Antigen | ULBP-1 |
|---|---|
| Clone | MM0592-10K10 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 1:50-1:100, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready |
| Applications | Flow Cytometry, Blocking Assay, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18001637
|
Novus Biologicals
NBP2-11986-0.025MG |
0.025 mg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18072277
|
Novus Biologicals
NBP2-11986 |
0.1 mg | |||||||
Description
ULBP-1 Monoclonal specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
| ULBP-1 | |
| LYOPH | |
| Flow Cytometry, Blocking Assay, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 | |
| ULBP1 | |
| Protein G purified |
| MM0592-10K10 | |
| Flow Cytometry 1:50-1:100, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9BZM6 | |
| 80329 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title