missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-1 Antibody (13C8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26263-100ul
This item is not returnable.
View return policy
Description
ULBP-1 Monoclonal antibody specifically detects ULBP-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ULBP-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 13C8 | |
| Western Blot 1:500-1:2000, ELISA | |
| ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 | |
| A synthesized peptide derived from Human ULBP-1 [UniProt Q9BZM6] | |
| 100 μL | |
| Immunology | |
| 80329 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction